Methyl-2-(brommethyl)-3-nitrobenzoat

Lieferant: CHEMPUR

Synonyme: Methyl 2-(bromomethyl)-3-nitrobenzoate

03-0004.1 03-0004.5
CHMP03-0004.1EA 0 EUR
CHMP03-0004.1 CHMP03-0004.5
Methyl-2-(brommethyl)-3-nitrobenzoat
Methyl-2-(brommethyl)-3-nitrobenzoat
Formel: C₉H₈BrNO₄
Molecular Weight: 274.07 g/mol
MDL: MFCD04114315
CAS-Nummer: 98475-07-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR