Methyl-2-(brommethyl)-3,5-dinitrobenzoat

Lieferant: CHEMPUR

03-0015.1 03-0015.5
CHMP03-0015.1EA 0 EUR
CHMP03-0015.1 CHMP03-0015.5
Methyl-2-(brommethyl)-3,5-dinitrobenzoat
Methyl-2-(brommethyl)-3,5-dinitrobenzoat
Formel: C₉H₇BrN₂O₆
MDL: MFCD04114324
CAS-Nummer: 153754-31-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR