1,6-Naphthyridin-2-carbonsäure

Lieferant: CHEMPUR

05-0045.1 05-0045.5
CHMP05-0045.1EA 0 EUR
CHMP05-0045.1 CHMP05-0045.5
1,6-Naphthyridin-2-carbonsäure
1,6-Naphthyridine-2-carboxylic acid
Formel: C₉H₆N₂O₂
Molecular Weight: 174.1600 g/mol
MDL: MFCD00202908
CAS-Nummer: 197507-59-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR