3-Nitrobenzoylacetonitril

Lieferant: CHEMPUR

Synonyme: 3-(3-Nitrophenyl)-3-oxopropanenitrile

05-0046.1 05-0046.5
CHMP05-0046.1EA 0 EUR
CHMP05-0046.1 CHMP05-0046.5
3-Nitrobenzoylacetonitril
3-Nitrobenzoylacetonitril
Formel: C₉H₆N₂O₃
Molecular Weight: 190.16
CAS-Nummer: 21667-64-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR