2-Brombenzoylacetonitril

Lieferant: CHEMPUR

05-0055.1 05-0055.5
CHMP05-0055.1EA 0 EUR
CHMP05-0055.1 CHMP05-0055.5
2-Brombenzoylacetonitril
2-Brombenzoylacetonitril
Formel: C₉H₆BrNO
Molecular Weight: 224.06 g/mol
CAS-Nummer: 53882-80-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR