2-Brom-4-fluorbenzaldehyd

Lieferant: CHEMPUR

10-0063.1 10-0063.5
CHMP10-0063.1EA 0 EUR
CHMP10-0063.1 CHMP10-0063.5
2-Brom-4-fluorbenzaldehyd
2-Bromo-4-fluorobenzaldehyde
Formel: C₇H₄BrFO
MDL: MFCD00672923
CAS-Nummer: 59142-68-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR