Certrichlorid Hydrat

Lieferant: CHEMPUR
Achtung

Synonyme: Cerium(III) chloride hydrate, Cerous chloride

1172.0250
CHMP1172.0250EA 0 EUR
CHMP1172.0250
Certrichlorid Hydrat
Certrichlorid Hydrat
Formel: CeCl₃·xH₂O
Molecular Weight: 246,48 g/mol (anhy)
MDL: MFCD00149633
CAS-Nummer: 19423-76-8
EINECS: 232-227-8
Merck Index: 12,02041

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR