2-Chlor-4-fluor-5-nitrophenol

Lieferant: BIONET RESEARCH

11L-319S.500 11L-319S.1 11L-319S.5 11L-319S.10 11L-319S.005
BIOT11L-319S.500EA 0 EUR
BIOT11L-319S.500 BIOT11L-319S.1 BIOT11L-319S.5 BIOT11L-319S.10 CHMP11L-319S.005
2-Chlor-4-fluor-5-nitrophenol
2-Chlor-4-fluor-5-nitrophenol
Formel: C₆H₃ClFNO₃
Molecular Weight: 191.5400 g/mol
MDL: MFCD02670258
CAS-Nummer: 84478-75-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR