3,5-Dichlorbenzamidin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: 3,5-Dichlorphenylamidin Hydrochlorid, 3,5-Dichlorbenzoesäureamidin Hydrochlorid

12-0035.500 12-0035.1
CHMP12-0035.500EA 0 EUR
CHMP12-0035.500 CHMP12-0035.1
3,5-Dichlorbenzamidin Hydrochlorid
3,5-Dichlorbenzamidin Hydrochlorid
Formel: C₇H₇Cl₃N₂
Molecular Weight: 225.5 g/mol
MDL: MFCD00173785
CAS-Nummer: 22978-61-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR