3-Methoxybenzamidin Hydrochlorid

Lieferant: CHEMPUR

12-0046.1 12-0046.5
CHMP12-0046.1EA 0 EUR
CHMP12-0046.1 CHMP12-0046.5
3-Methoxybenzamidin Hydrochlorid
3-Methoxybenzamidin Hydrochlorid
Formel: C₈H₁₁ClN₂O
Molecular Weight: 186.6400 g/mol
MDL: MFCD04114430
CAS-Nummer: 26113-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR