2,3,4,5,6-Pentafluorbenzamidin Hydrochlorid

Lieferant: CHEMPUR

12-0048.1 12-0048.5
CHMP12-0048.1EA 0 EUR
CHMP12-0048.1 CHMP12-0048.5
2,3,4,5,6-Pentafluorbenzamidin Hydrochlorid
2,3,4,5,6-Pentafluorbenzamidin Hydrochlorid
Formel: C₇H₄ClF₅N₂
MDL: MFCD04114451
CAS-Nummer: 885954-06-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR