2,3-Difluorbenzamidin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: 2,3-Difluorbenzoesäureamidin Hydrochlorid, 2,3-Difluorphenylamidin Hydrochlorid

12-0061.500 12-0061.1
CHMP12-0061.500EA 0 EUR
CHMP12-0061.500 CHMP12-0061.1
2,3-Difluorbenzamidin Hydrochlorid
2,3-Difluorbenzamidin Hydrochlorid
Formel: C₇H₇ClF₂N₂
Molecular Weight: 192.6 g/mol
MDL: MFCD04114444
CAS-Nummer: 1138036-26-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR