3,5-Difluorbenzamidin Hydrochlorid
Lieferant: CHEMPUR
Synonyme:
3,5-Difluorphenylamidin Hydrochlorid, 3,5-Difluorbenzoesäureamidin Hydrochlorid
12-0066.500
12-0066.1
CHMP12-0066.500EA
0
EUR
CHMP12-0066.500
CHMP12-0066.1
3,5-Difluorbenzamidin Hydrochlorid
3,5-Difluorbenzamidin Hydrochlorid
Formel:
C₇H₇ClF₂N₂ Molecular Weight: 192.6 g/mol |
MDL:
MFCD04114449 CAS-Nummer: 144797-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...