3,5-Difluorbenzamidin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: 3,5-Difluorphenylamidin Hydrochlorid, 3,5-Difluorbenzoesäureamidin Hydrochlorid

12-0066.500 12-0066.1
CHMP12-0066.500EA 0 EUR
CHMP12-0066.500 CHMP12-0066.1
3,5-Difluorbenzamidin Hydrochlorid
3,5-Difluorbenzamidin Hydrochlorid
Formel: C₇H₇ClF₂N₂
Molecular Weight: 192.6 g/mol
MDL: MFCD04114449
CAS-Nummer: 144797-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR