2,4-Dimethylbenzylbromid

Lieferant: CHEMPUR

Synonyme: 1-(Bromomethyl)-2,4-dimethylbenzene

221500-5G
CHMP221500-5GEA 0 EUR
CHMP221500-5G
2,4-Dimethylbenzylbromid
2,4-Dimethylbenzylbromid
Formel: C₉H₁₁Br
Molecular Weight: 199,09 g/mol
MDL: MFCD03844740
CAS-Nummer: 78831-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR