Isochinolin-6-amin

Lieferant: CHEMPUR

Synonyme: 6-Aminoisoquinoline

50-0055.1 50-0055.5
CHMP50-0055.1EA 0 EUR
CHMP50-0055.1 CHMP50-0055.5
Isochinolin-6-amin
Isochinolin-6-amin
Formel: C₉H₈N₂
MDL: MFCD04114862
CAS-Nummer: 23687-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR