1-Boc-4-chlorpiperidin

Lieferant: CHEMPUR

Synonyme: tert-Butyl 4-chloropiperidine-1-carboxylate

60-0170.1 60-0170.5
CHMP60-0170.1EA 0 EUR
CHMP60-0170.1 CHMP60-0170.5
1-Boc-4-chlorpiperidin
1-Boc-4-chlorpiperidin
Formel: C₁₀H₁₈ClNO₂
MDL: MFCD04115040
CAS-Nummer: 154874-94-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR