Pyrazolo[1,5-a]pyridin-2-carbonsäureamid

Lieferant: CHEMPUR

68-0053.1 68-0053.500
CHMP68-0053.1EA 0 EUR
CHMP68-0053.1 CHMP68-0053.500
Pyrazolo[1,5-a]pyridin-2-carbonsäureamid
Pyrazolo[1,5-a]pyridin-2-carbonsäureamid
Formel: C₈H₇N₃O
Molecular Weight: 161.1607 g/mol
MDL: MFCD04115109
CAS-Nummer: 885275-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR