4-Brom-2-methylpyridin

Lieferant: CHEMPUR

69-0011.1 69-0011.5
CHMP69-0011.1EA 0 EUR
CHMP69-0011.1 CHMP69-0011.5
4-Brom-2-methylpyridin
4-Brom-2-methylpyridin
Formel: C₆H₆BrN
Molecular Weight: 172.02 g/mol
MDL: MFCD03788196
CAS-Nummer: 22282-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR