1-Boc-piperidin-3-carbonsäure

Lieferant: CHEMPUR

Synonyme: N-Boc-DL-nipecotic acid, 1-[(tert-butoxy)carbonyl]piperidine-3-carboxylic acid, 1-Boc-Nipecotic acid, Boc-Nip-OH, 1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid, N-BOC-3-Piperidinecarboxylic acid

75-0030.1 75-0030.5
CHMP75-0030.1EA 0 EUR
CHMP75-0030.1 CHMP75-0030.5
1-Boc-piperidin-3-carbonsäure
1-Boc-piperidin-3-carbonsäure
Formel: C₁₁H₁₉NO₄
MDL: MFCD00673775
CAS-Nummer: 84358-12-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR