4-Chlor-7-fluor-2-phenylchinazolin

Lieferant: CHEMPUR

79-0010.500 79-0010.1
CHMP79-0010.500EA 0 EUR
CHMP79-0010.500 CHMP79-0010.1
4-Chlor-7-fluor-2-phenylchinazolin
4-Chloro-7-fluoro-2-phenylquinazoline
Formel: C₁₄H₈ClFN₂
MDL: MFCD04115228
CAS-Nummer: 885277-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR