3-(Dimethylamino)phenol
Lieferant: CHEMPUR
Synonyme:
N,N-Dimethyl-3-hydroxyanilin
80296
CHMP80296EA
0
EUR
CHMP80296
3-(Dimethylamino)phenol
3-(Dimethylamino)phenol
Formel:
C₈H₁₁NO Molecular Weight: 137,18 g/mol Siedepunkt: 265 °C (1013 hPa) Schmelzpunkt: 65…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002264 CAS-Nummer: 99-07-0 EINECS: 202-727-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...