2,4-Dibenzyloxy-5-pyrimidinboronsäure
Lieferant: CHEMPUR
Synonyme:
2,4-Bis(benzyloxy)pyrimidine-5-boronic acid, 2,4-Bis(benzyloxy)pyrimidin-5-ylboronic acid
902424.POR
CHMP902424.POREA
0
EUR
CHMP902424.POR
2,4-Dibenzyloxy-5-pyrimidinboronsäure
2,4-Dibenzyloxy-5-pyrimidinboronsäure
Formel:
C₁₈H₁₇BN₂O₄ Molecular Weight: 336,150 g/mol |
MDL:
MFCD01318938 CAS-Nummer: 70523-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...