2,3,4-Trimethoxyphenylboronsäure 95+%

Lieferant: CHEMPUR

Synonyme: (2,3,4-Trimethoxyphenyl)metaborsäure, 2,3,4-Trimethoxybenzolboronsäure, Dihydroxy(2,3,4-trimethoxyphenyl)boran

902615.POR
CHMP902615.POREA 0 EUR
CHMP902615.POR
2,3,4-Trimethoxyphenylboronsäure 95+%
2,3,4-Trimethoxyphenylboronsäure
Formel: C₉H₁₃BO₅
Molecular Weight: 212,01 g/mol
MDL: MFCD01318183
CAS-Nummer: 118062-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR