7-Nitro-1H-indazol
Lieferant: CHEMPUR
Synonyme:
7-NINA, 7-Nitro-1H-indazol
AC32877.0.1
CHMPAC32877.0.1EA
0
EUR
CHMPAC32877.0.1
7-Nitro-1H-indazol
7-Nitro-1H-indazole
Formel:
C₇H₅N₃O₂ Molecular Weight: 163,14 g/mol Schmelzpunkt: 190 °C Dichte: 1,56 g/cm³ (293 °C) |
MDL:
MFCD00022789 CAS-Nummer: 2942-42-9 EINECS: 220-934-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...