5-Formyl-2-methoxyphenylboronsäure ≥97%
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
5-Formyl-2-methoxybenzeneboronic acid, (5-Formyl-2-methoxyphenyl)boronic acid
AC35954.0.1
CHMPAC35954.0.1EA
0
EUR
CHMPAC35954.0.1
5-Formyl-2-methoxyphenylboronsäure ≥97%
5-Formyl-2-methoxyphenylboronsäure
Formel:
C₈H₉BO₄ Lagertemperatur: Kühlschrank |
MDL:
MFCD01319044 CAS-Nummer: 127972-02-5 |
Spezifikation Testergebnisse
Appearance | Light orange Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...