3-(Methylthio)phenylboronsäure

Lieferant: CHEMPUR

Synonyme: 3-Thioanisoleboronic acid, 3-(Methylthio)benzeneboronic acid

AC35970.0.1 AC35970.250
CHMPAC35970.0.1EA 0 EUR
CHMPAC35970.0.1 MAYBAC35970.250
3-(Methylthio)phenylboronsäure
3-(Methylthio)phenylboronsäure
Formel: C₇H₉BO₂S
Molecular Weight: 168.02 g/mol
MDL: MFCD01319105
CAS-Nummer: 128312-11-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR