Ethyl-2-amino-5-methyl-4-phenyl-3-thiophencarboxylat

Lieferant: CHEMPUR

AW00937.250 AW00937DA
CHMPAW00937.250EA 0 EUR
CHMPAW00937.250 MAYBAW00937DA
Ethyl-2-amino-5-methyl-4-phenyl-3-thiophencarboxylat
Ethyl-2-amino-5-methyl-4-phenyl-3-thiophencarboxylat
Formel: C₁₄H₁₅NO₂S
Molecular Weight: 261,34 g/mol
MDL: MFCD01763475
CAS-Nummer: 4815-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR