3-Brom-5-nitrosalicylaldehyd

Lieferant: CHEMPUR

Synonyme: 3-Brom-2-hydroxy-5-nitrobenzaldehyd

BTB03755.250 BTB03755DA
CHMPBTB03755.250EA 0 EUR
CHMPBTB03755.250 MAYBBTB03755DA
3-Brom-5-nitrosalicylaldehyd
3-Brom-5-nitrosalicylaldehyd
Formel: C₇H₄BrNO₄
Molecular Weight: 246,02 g/mol
MDL: MFCD00051833
CAS-Nummer: 16789-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR