2,4-Dichlor-6-nitrophenol
Lieferant: CHEMPUR
|
Achtung
|
BTB04020.250
BTB04020.1
CHMPBTB04020.250EA
0
EUR
CHMPBTB04020.250
MAYBBTB04020.1
2,4-Dichlor-6-nitrophenol
2,4-Dichlor-6-nitrophenol
Formel:
C₆H₃Cl₂NO₃ Molecular Weight: 208 g/mol Siedepunkt: 243 Schmelzpunkt: 105 |
MDL:
MFCD00007101 CAS-Nummer: 609-89-2 EINECS: 210-202-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...