5-Nitrosalicylaldehyd
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
2-Hydroxy-5-nitrobenzaldehyde
BTB04054.250
BTB04054.1
CHMPBTB04054.250EA
0
EUR
CHMPBTB04054.250
MAYBBTB04054.1
5-Nitrosalicylaldehyd
5-Nitrosalicylaldehyd
Formel:
C₇H₅NO₄ Molecular Weight: 167.12 g/mol Schmelzpunkt: 123…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007337 CAS-Nummer: 97-51-8 EINECS: 202-587-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...