5-Fluor-4-chinazolon

Lieferant: CHEMPUR

Synonyme: 5-Fluoroquinazolin-4(1H)-one, 5-Fluoro-4-hydroxyquinazoline

BTB04061.10
CHMPBTB04061.10EA 0 EUR
CHMPBTB04061.10
5-Fluor-4-chinazolon
5-Fluor-4-chinazolon
Formel: C₈H₅FN₂O
Molecular Weight: 164,13 g/mol
MDL: MFCD09475836
CAS-Nummer: 436-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR