2-Chlornicotinoylchlorid

Lieferant: CHEMPUR

BTB06401.250
CHMPBTB06401.250EA 0 EUR
CHMPBTB06401.250
2-Chlornicotinoylchlorid
2-Chlornicotinoylchlorid
Formel: C₆H₃Cl₂NO
Molecular Weight: 176,00 g/mol
MDL: MFCD00051677
CAS-Nummer: 49609-84-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR