2-Amino-3,5-dimethylpyrazin

Lieferant: CHEMPUR

Synonyme: 3,5-Dimethylpyrazin-2-amine

BTB07911.250
CHMPBTB07911.250EA 0 EUR
CHMPBTB07911.250
2-Amino-3,5-dimethylpyrazin
2-Amino-3,5-dimethylpyrazine
Formel: C₆H₉N₃
Molecular Weight: 123,1558 g/mol
MDL: MFCD00107172
CAS-Nummer: 91678-81-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR