4-[(3,4-Dichlorbenzyl)oxy]benzaldehyd

Lieferant: CHEMPUR

BTB09518.250 BTB09518DA
CHMPBTB09518.250EA 0 EUR
CHMPBTB09518.250 MAYBBTB09518DA
4-[(3,4-Dichlorbenzyl)oxy]benzaldehyd
4-[(3,4-Dichlorbenzyl)oxy]benzaldehyd
Formel: C₁₄H₁₀Cl₂O₂
MDL: MFCD00173652
CAS-Nummer: 66742-56-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR