4'-Amino-2'-hydroxyacetophenon

Lieferant: CHEMPUR

Synonyme: 1-(4-Amino-2-hydroxyphenyl)ethanone, 1-(4-Amino-2-hydroxyphenyl)ethan-1-one

BTB10056.250
CHMPBTB10056.250EA 0 EUR
CHMPBTB10056.250
4'-Amino-2'-hydroxyacetophenon
4'-Amino-2'-hydroxyacetophenon
Formel: C₈H₉NO₂
Molecular Weight: 151,1600 g/mol
MDL: MFCD00100636
CAS-Nummer: 2476-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR