7-Methoxy-3,7-dimethyl-1-octen

Lieferant: CHEMPUR

BTB12333.250 BTB12333.1
CHMPBTB12333.250EA 0 EUR
CHMPBTB12333.250 MAYBBTB12333.1
7-Methoxy-3,7-dimethyl-1-octen
7-Methoxy-3,7-dimethyl-1-octen
Formel: C₁₁H₂₂O
Molecular Weight: 170.3 g/mol
CAS-Nummer: 53767-86-5
EINECS: 258-750-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR