Methylpicolinat

Lieferant: CHEMPUR

BTB14002.250 BTB14002.1
CHMPBTB14002.250EA 0 EUR
CHMPBTB14002.250 MAYBBTB14002.1
Methylpicolinat
Methylpicolinat
Formel: C₇H₇NO₂
MDL: MFCD00038038
CAS-Nummer: 2459-07-6
EINECS: 219-545-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR