6-Bromchromon

Lieferant: CHEMPUR

BTB14078.250 BTB14078.1
CHMPBTB14078.250EA 0 EUR
CHMPBTB14078.250 MAYBBTB14078.1
6-Bromchromon
6-Bromchromon
Formel: C₉H₅BrO₂
MDL: MFCD00239369
CAS-Nummer: 51483-92-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR