3-Methylbenzoesäureanhydrid

Lieferant: CHEMPUR

BTB14938.250 BTB14938.1
CHMPBTB14938.250EA 0 EUR
CHMPBTB14938.250 MAYBBTB14938.1
3-Methylbenzoesäureanhydrid
3-Methylbenzoic anhydride
Formel: C₁₆H₁₄O₃
MDL: MFCD00048083
CAS-Nummer: 21436-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR