Pentabrompyridin

Lieferant: CHEMPUR

BTB15039.250 BTB15039.1
CHMPBTB15039.250EA 0 EUR
CHMPBTB15039.250 MAYBBTB15039.1
Pentabrompyridin
Pentabrompyridin
Formel: C₅Br₅N
CAS-Nummer: 2402-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR