5-Chlorpentylacetat
Lieferant: CHEMPUR
Synonyme:
Essigsäure(5-chlorpentyl)ester, Essigsäure(5-chloramyl)ester, 5-Chloramylacetat
BTBS00044.250
CHMPBTBS00044.250EA
0
EUR
CHMPBTBS00044.250
5-Chlorpentylacetat
5-Chlorpentylacetat
Formel:
C₇H₁₃ClO₂ Molecular Weight: 164.63 g/mol Siedepunkt: 101…104 °C (1013 hPa) Dichte: 1.0648 g/cm³ (20 °C) |
MDL:
MFCD00013696 CAS-Nummer: 20395-28-2 EINECS: 243-784-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...