[5-Methyl-3-phenyl-4-isoxazolyl]methanol

Lieferant: CHEMPUR

CC00509.250
CHMPCC00509.250EA 0 EUR
CHMPCC00509.250
[5-Methyl-3-phenyl-4-isoxazolyl]methanol
[5-Methyl-3-phenyl-4-isoxazolyl]methanol
Formel: C₁₁H₁₁NO₂
MDL: MFCD02677724
CAS-Nummer: 18718-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR