3,5-Dimethyl-4-isoxazolcarbonsäure

Lieferant: CHEMPUR

Synonyme: 3,5-Dimethylisoxasole-4-carboxylic acid

CC00601.250 CC00601DA
CHMPCC00601.250EA 0 EUR
CHMPCC00601.250 MAYBCC00601DA
3,5-Dimethyl-4-isoxazolcarbonsäure
3,5-Dimethylisoxazole-4-carboxylic acid
Formel: C₆H₇NO₃
Molecular Weight: 141,13 g/mol
MDL: MFCD00051657
CAS-Nummer: 2510-36-3
EINECS: 219-724-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR