3,5-Dimethyl-4-iod-1H-pyrazol

Lieferant: CHEMPUR

Synonyme: 3,5-Dimethyl-4-iodopyrazole

CC01211.250
CHMPCC01211.250EA 0 EUR
CHMPCC01211.250
3,5-Dimethyl-4-iod-1H-pyrazol
3,5-Dimethyl-4-iodo-1H-pyrazole
Formel: C₅H₇IN₂
Molecular Weight: 222,03 g/mol
MDL: MFCD00040247
CAS-Nummer: 2033-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR