1,3-Benzodioxol-4-carbaldehyd

Lieferant: CHEMPUR

Synonyme: Benzo[d][1,3]dioxole-4-carbaldehyde, 2,3-(Methylenedioxy)benzaldehyde, 1,3-Benzodioxole-4-carboxaldehyde

CC01704.250
CHMPCC01704.250EA 0 EUR
CHMPCC01704.250
1,3-Benzodioxol-4-carbaldehyd
1,3-Benzodioxol-4-carbaldehyd
Formel: C₈H₆O₃
Molecular Weight: 150,1300 g/mol
MDL: MFCD00016902
CAS-Nummer: 7797-83-3
EINECS: 232-252-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR