1-Methylpyrrol-2-carbonsäure

Lieferant: CHEMPUR

Synonyme: 1-Methyl-1H-pyrrole-2-carboxylic acid, 1-Methyl-2-pyrrolecarboxylic acid

CC02701.250
CHMPCC02701.250EA 0 EUR
CHMPCC02701.250
1-Methylpyrrol-2-carbonsäure
1-Methylpyrrole-2-carboxylic acid
Formel: C₆H₇NO₂
Molecular Weight: 125.13 g/mol
MDL: MFCD00003088
CAS-Nummer: 6973-60-0
EINECS: 230-217-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR