2-Chinolincarbaldehyd

Lieferant: CHEMPUR
Achtung

Synonyme: 2-Chinolincarboxaldehyd

CC04404.250 CC04404DA
CHMPCC04404.250EA 0 EUR
CHMPCC04404.250 MAYBCC04404DA
2-Chinolincarbaldehyd
2-Chinolincarbaldehyd
Formel: C₁₀H₇NO
Molecular Weight: 157,17 g/mol
MDL: MFCD00075032
CAS-Nummer: 5470-96-2
EINECS: 226-804-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR