4-Chinolincarbonsäure

Lieferant: CHEMPUR
Achtung

Synonyme: Chinolin-4-carbonsäure

CC04601.250 CC04601DA
CHMPCC04601.250EA 0 EUR
CHMPCC04601.250 MAYBCC04601DA
4-Chinolincarbonsäure
4-Chinolincarbonsäure
Formel: C₁₀H₇NO₂
Molecular Weight: 173,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006782
CAS-Nummer: 486-74-8
EINECS: 207-640-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR