6-Brombenzothiazol

Lieferant: CHEMPUR

Synonyme: 6-Bromobenzo[d]thiazole

CC05910DA CC05910.10
MAYBCC05910DAEA 0 EUR
MAYBCC05910DA CHMPCC05910.10
6-Brombenzothiazol
6-Bromobenzothiazole
Formel: C₇H₄BrNS
Molecular Weight: 214,09 g/mol
MDL: MFCD04115372
CAS-Nummer: 53218-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR